Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WEE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143976.1 | NP_001137448.1 | 2 | 871 | ||
NM_003390.3 | NP_003381.1 | 2 | 1011 | |||
GenBank mRNA | AK122837.1 | - | 2 | 871 | ||
BC051831.1 | - | 2 | 809 | 6147695 | ||
BC070052.1 | - | 2 | 1001 | 30336707 | ||
BX641032.1 | - | 1 | 529 | |||
U10564.1 | - | 2 | 1011 | |||
X62048.1 | - | 2 | 928 |