Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011658.3 | NP_001011658.1 | 4 | 441 | ||
NM_001128835.2 | NP_001122307.2 | 4 | 435 | |||
NM_014563.5 | NP_055378.1 | 3 | 299 | |||
XM_011545565.1 | XP_011543867.1 | 5 | 509 | |||
XM_011545566.2 | XP_011543868.1 | 4 | 365 | |||
GenBank mRNA | AK098253.1 | - | 2 | 2448 | ||
AK310542.1 | - | 4 | 345 | |||
AK313736.1 | - | 4 | 381 | |||
BC016915.1 | - | 3 | 262 | 4432196 | ||
BC052618.1 | - | 3 | 262 | 6581382 | ||
BI821518.1 | - | 3 | 273 | |||
CB962763.1 | - | 3 | 284 | |||
CD698535.1 | - | 4 | 395 | |||
DA344866.1 | - | 3 | 299 | |||
DA350753.1 | - | 4 | 429 | |||
DA542477.1 | - | 4 | 375 |