Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098811.1 | NP_001092281.1 | 5 | 923 | ||
NM_001098812.1 | NP_001092282.1 | 5 | 923 | |||
NM_001098813.1 | NP_001092283.1 | 5 | 849 | |||
NM_001300798.1 | NP_001287727.1 | 5 | 917 | |||
NM_001300799.1 | NP_001287728.1 | 5 | 917 | |||
NM_015146.1 | NP_055961.1 | 5 | 923 | |||
XM_005271927.2 | XP_005271984.1 | 5 | 925 | |||
XM_005271928.2 | XP_005271985.1 | 5 | 919 | |||
XM_005271931.3 | XP_005271988.1 | 6 | 1006 | |||
XM_011543288.1 | XP_011541590.1 | 5 | 945 | |||
XM_017009254.1 | XP_016864743.1 | 5 | 919 | |||
XM_017009255.1 | XP_016864744.1 | 5 | 992 | |||
XM_017009256.1 | XP_016864745.1 | 5 | 986 | |||
XM_017009257.1 | XP_016864746.1 | 5 | 939 | |||
XM_017009258.1 | XP_016864747.1 | 5 | 992 | |||
XM_017009259.1 | XP_016864748.1 | 5 | 986 | |||
XM_017009260.1 | XP_016864749.1 | 5 | 939 | |||
XM_017009261.1 | XP_016864750.1 | 5 | 986 | |||
GenBank mRNA | AF179995.1 | - | 5 | 923 | ||
AF440761.1 | - | 5 | 923 | |||
AF440762.1 | - | 5 | 923 | |||
AF440763.1 | - | 5 | 758 | |||
AK057797.1 | - | 5 | 781 | |||
BC001329.1 | - | 2 | 322 | |||
BC140759.1 | - | 5 | 739 | 9021627 | ||
BC171779.1 | - | 5 | 739 | 9054433 | ||
D86957.1 | - | 5 | 923 |