Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001665.3 | NP_001656.2 | 2 | 946 | ||
XM_005252916.2 | XP_005252973.1 | 2 | 1126 | |||
XM_017017719.1 | XP_016873208.1 | 2 | 973 | |||
GenBank mRNA | BC104177.2 | - | 1 | 871 | 40030689 | |
BC104178.2 | - | 2 | 873 | 40030690 | ||
BX359696.2 | - | 1 | 859 | |||
X61587.1 | - | 2 | 915 |