Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135729.1 | NP_001129201.1 | 8 | 921 | ||
NM_001135730.1 | NP_001129202.1 | 7 | 859 | |||
NM_001135732.1 | NP_001129204.1 | 8 | 994 | |||
NM_005488.2 | NP_005479.1 | 8 | 994 | |||
NR_024194.1 | 8 | 1023 | ||||
NR_024195.1 | 7 | 944 | ||||
XM_011529818.2 | XP_011528120.1 | 9 | 1008 | |||
XM_011529820.2 | XP_011528122.1 | 7 | 859 | |||
XM_017028529.1 | XP_016884018.1 | 9 | 1008 | |||
XM_017028530.1 | XP_016884019.1 | 7 | 859 | |||
GenBank mRNA | AJ006973.1 | - | 8 | 930 | ||
AK026576.1 | - | 8 | 947 | |||
AK097066.1 | - | 6 | 505 | |||
AK097926.1 | - | 9 | 1010 | |||
AK293694.1 | - | 8 | 921 | |||
AK295918.1 | - | 7 | 854 | |||
AK296670.1 | - | 8 | 931 | |||
AK297827.1 | - | 7 | 773 | |||
AK314522.1 | - | 8 | 903 | |||
BC040006.1 | - | 8 | 887 | |||
BC046151.1 | - | 8 | 872 | 6470390 | ||
CR456595.1 | - | 8 | 890 |