Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135771.2 | NP_001129243.1 | 11 | 1654 | ||
NM_001324299.1 | NP_001311228.1 | 12 | 1750 | |||
NM_001324301.1 | NP_001311230.1 | 13 | 1798 | |||
NM_001324302.1 | NP_001311231.1 | 12 | 1750 | |||
NM_001324303.1 | NP_001311232.1 | 12 | 1750 | |||
NM_001324304.1 | NP_001311233.1 | 12 | 1750 | |||
NM_001324305.1 | NP_001311234.1 | 13 | 1798 | |||
NM_001324306.1 | NP_001311235.1 | 10 | 1478 | |||
NM_002951.4 | NP_002942.2 | 12 | 1750 | |||
XM_006723851.2 | XP_006723914.1 | 13 | 1603 | |||
XM_006723852.2 | XP_006723915.1 | 13 | 1603 | |||
GenBank mRNA | AK096243.1 | - | 11 | 1539 | ||
AK296126.1 | - | 10 | 1153 | |||
AK308683.1 | - | 10 | 1177 | |||
AK315801.1 | - | 12 | 1478 | |||
BC002380.2 | - | 9 | 1024 | |||
BC003560.1 | - | 12 | 1476 | 3546673 | ||
BC013028.2 | - | 4 | 390 | |||
BC020222.1 | - | 12 | 1621 | 3640766 | ||
CR456899.1 | - | 12 | 1439 | |||
Y00282.1 | - | 12 | 1727 |