Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135771.2 | NP_001129243.1 | 14 | 1944 | ||
NM_001324299.1 | NP_001311228.1 | 14 | 1944 | |||
NM_001324301.1 | NP_001311230.1 | 16 | 2088 | |||
NM_001324302.1 | NP_001311231.1 | 14 | 1944 | |||
NM_001324303.1 | NP_001311232.1 | 15 | 2040 | |||
NM_001324304.1 | NP_001311233.1 | 15 | 2040 | |||
NM_001324305.1 | NP_001311234.1 | 16 | 2088 | |||
NM_001324306.1 | NP_001311235.1 | 13 | 1768 | |||
NM_002951.4 | NP_002942.2 | 15 | 2040 | |||
XM_006723851.2 | XP_006723914.1 | 15 | 1797 | |||
XM_006723852.2 | XP_006723915.1 | 15 | 1797 | |||
GenBank mRNA | AK096243.1 | - | 14 | 1829 | ||
AK315801.1 | - | 15 | 1768 | |||
BC002380.2 | - | 12 | 1314 | |||
BC003560.1 | - | 15 | 1766 | 3546673 | ||
BC013028.2 | - | 7 | 680 | |||
BC020222.1 | - | 15 | 1911 | 3640766 | ||
BF923732.1 | - | 2 | 111 | |||
CR456899.1 | - | 15 | 1729 | |||
Y00282.1 | - | 15 | 2017 |