Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000404.3 | NP_000395.2 | 12 | 1352 | ||
NM_001079811.2 | NP_001073279.1 | 12 | 1246 | |||
NM_001135602.2 | NP_001129074.1 | 9 | 959 | |||
NM_001317040.1 | NP_001303969.1 | 13 | 1496 | |||
GenBank mRNA | AK130133.1 | - | 12 | 1280 | ||
AK222763.1 | - | 12 | 1243 | |||
AK223091.1 | - | 12 | 1268 | |||
AK298985.1 | - | 9 | 871 | |||
AK300021.1 | - | 12 | 1179 | |||
AK300753.1 | - | 13 | 1412 | |||
AK312988.1 | - | 12 | 1239 | |||
BC007493.2 | - | 12 | 1254 | 2988086 | ||
BT007147.1 | - | 12 | 1207 | |||
M22590.1 | - | 12 | 1241 | |||
M27507.1 | - | 12 | 1257 | |||
M27508.1 | - | 9 | 864 | |||
M34423.1 | - | 12 | 1267 |