Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319243.1 | NP_001306172.1 | 8 | 1745 | ||
NM_001319244.1 | NP_001306173.1 | 7 | 1206 | |||
NM_001319302.1 | NP_001306231.1 | 8 | 2156 | |||
NM_178314.4 | NP_847884.2 | 7 | 1657 | |||
XM_006719372.2 | XP_006719435.1 | 8 | 1675 | |||
XM_006719373.3 | XP_006719436.1 | 7 | 1028 | |||
XM_011538282.2 | XP_011536584.1 | 8 | 1716 | |||
XM_011538284.2 | XP_011536586.1 | 7 | 1247 | |||
XM_011538285.2 | XP_011536587.1 | 7 | 1008 | |||
GenBank mRNA | AK096697.1 | - | 9 | 1587 | ||
BC041375.1 | - | 1 | 3763 | |||
BC080626.1 | - | 7 | 1206 | 6339887 | ||
BC089444.1 | - | 7 | 1459 | 30559974 |