Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114357.1 | NP_001107829.1 | 6 | 760 | ||
XM_005263019.3 | XP_005263076.1 | 8 | 1113 | |||
XM_011531989.2 | XP_011530291.1 | 7 | 1039 | |||
XM_017008236.1 | XP_016863725.1 | 8 | 2506 | |||
XM_017008237.1 | XP_016863726.1 | 9 | 1281 | |||
XM_017008238.1 | XP_016863727.1 | 7 | 2290 | |||
XM_017008239.1 | XP_016863728.1 | 7 | 2293 | |||
XM_017008240.1 | XP_016863729.1 | 7 | 1042 | |||
XM_017008241.1 | XP_016863730.1 | 6 | 2268 | |||
GenBank mRNA | BC127739.1 | - | 4 | 465 | ||
BC141968.1 | - | 4 | 464 | |||
BC150574.1 | - | 6 | 760 | 40127870 |