Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015072.4 | NP_055887.3 | 26, 27 | 3266 | ||
GenBank mRNA | AB023215.1 | - | 24, 25 | 2826 | ||
AL136808.1 | - | 12, 13 | 1714 | |||
AY237126.1 | - | 26, 27 | 3196 | |||
BC016625.1 | - | 13, 14 | 1847 | |||
BC136472.1 | - | 26, 27 | 3156 | 9020461 | ||
BC136473.1 | - | 26, 27 | 3156 | 9020462 | ||
CR533554.1 | - | 11, 12 | 1644 |