Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM221A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127364.2 | NP_001120836.1 | 6 | 849 | ||
NM_001127365.2 | NP_001120837.1 | 5 | 675 | |||
NM_001300932.1 | NP_001287861.1 | 6 | 783 | |||
NM_199136.4 | NP_954587.2 | 7 | 957 | |||
XM_011515369.2 | XP_011513671.1 | 7 | 952 | |||
XM_011515370.2 | XP_011513672.1 | 7 | 949 | |||
XM_011515371.2 | XP_011513673.1 | 6 | 841 | |||
XM_011515372.2 | XP_011513674.1 | 7 | 1097 | |||
XM_017012134.1 | XP_016867623.1 | 6 | 844 | |||
XM_017012135.1 | XP_016867624.1 | 6 | 989 | |||
GenBank mRNA | AK127774.1 | - | 8 | 3569 | ||
AL832127.1 | - | 8 | 3626 | |||
BC026000.1 | - | 5 | 607 | |||
BC042034.1 | - | 6 | 830 | |||
BC063130.1 | - | 7 | 871 | 30382529 |