Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM221A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127364.2 | NP_001120836.1 | 3 | 362 | ||
NM_001127365.2 | NP_001120837.1 | 2 | 188 | |||
NM_001300932.1 | NP_001287861.1 | 2 | 188 | |||
NM_199136.4 | NP_954587.2 | 3 | 362 | |||
XM_011515369.2 | XP_011513671.1 | 3 | 357 | |||
XM_011515370.2 | XP_011513672.1 | 3 | 357 | |||
XM_011515371.2 | XP_011513673.1 | 3 | 357 | |||
XM_011515372.2 | XP_011513674.1 | 3 | 502 | |||
XM_017012134.1 | XP_016867623.1 | 3 | 357 | |||
XM_017012135.1 | XP_016867624.1 | 3 | 502 | |||
GenBank mRNA | AK127774.1 | - | 4 | 3011 | ||
AL832127.1 | - | 3 | 298 | |||
BC026000.1 | - | 2 | 120 | |||
BC042034.1 | - | 3 | 344 | |||
BC063130.1 | - | 3 | 276 | 30382529 | ||
BI549491.1 | - | 3 | 349 | |||
BU958830.1 | - | 2 | 120 | |||
CA867596.1 | - | 3 | 323 | |||
HY056255.1 | - | 3 | 363 |