Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135241.2 | NP_001128713.1 | 4 | 650 | ||
GenBank mRNA | AK091194.1 | - | 5 | 955 | ||
AK296686.1 | - | 4 | 497 |