Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102445.2 | NP_001095915.1 | 4 | 816 | ||
NM_001113380.1 | NP_001106851.1 | 3 | 391 | |||
NM_001113381.1 | NP_001106852.1 | 3 | 427 | |||
NM_005613.5 | NP_005604.1 | 3 | 707 | |||
GenBank mRNA | AB209019.1 | - | 1 | 1496 | ||
AF493928.1 | - | 3 | 196 | |||
AK093959.1 | - | 3 | 320 | |||
AK096204.1 | - | 2 | 623 | |||
AK295540.1 | - | 4 | 551 | |||
AK296240.1 | - | 3 | 235 | |||
AK312250.1 | - | 3 | 319 | |||
BC000737.1 | - | 3 | 281 | 3510260 | ||
BC051869.1 | - | 3 | 293 | 5786695 | ||
BI547160.1 | - | 3 | 421 | |||
BT007025.1 | - | 3 | 196 | |||
DA253378.1 | - | 4 | 551 | |||
DA660018.1 | - | 3 | 476 | |||
DA660695.1 | - | 3 | 463 | |||
DQ346662.1 | - | 3 | 320 | |||
DQ346663.1 | - | 3 | 535 | |||
DQ346664.1 | - | 4 | 647 | |||
EF054877.1 | - | 3 | 391 | |||
U27768.1 | - | 3 | 293 |