Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIPA1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015073.2 | NP_055888.1 | 3 | 1734 | ||
XM_005258671.4 | XP_005258728.1 | 3 | 2175 | |||
XM_011526657.2 | XP_011524959.1 | 3 | 2529 | |||
XM_017026517.1 | XP_016882006.1 | 4 | 2629 | |||
XM_017026518.1 | XP_016882007.1 | 3 | 2131 | |||
GenBank mRNA | AY168880.1 | - | 1 | 1480 | ||
BC150620.1 | - | 3 | 1713 | 9056990 |