Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098503.1 | NP_001091973.1 | 8 | 1910 | ||
NM_002843.3 | NP_002834.3 | 8 | 1910 | |||
XM_017018083.1 | XP_016873572.1 | 7 | 1641 | |||
XM_017018084.1 | XP_016873573.1 | 8 | 2023 | |||
XM_017018085.1 | XP_016873574.1 | 8 | 1522 | |||
GenBank mRNA | BC063417.1 | - | 8 | 1666 | ||
D37781.1 | - | 8 | 1899 | |||
U10886.1 | - | 8 | 1904 |