Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIPA1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015073.2 | NP_055888.1 | 7 | 2584 | ||
XM_005258671.4 | XP_005258728.1 | 7 | 3025 | |||
XM_011526657.2 | XP_011524959.1 | 7 | 3379 | |||
XM_017026517.1 | XP_016882006.1 | 8 | 3479 | |||
XM_017026518.1 | XP_016882007.1 | 7 | 2981 | |||
GenBank mRNA | AB011117.2 | - | 5 | 830 | ||
AY168880.1 | - | 5 | 2330 | |||
BC150620.1 | - | 7 | 2563 | 9056990 |