Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170747.1 | NP_001164218.1 | 3 | 325 | ||
NM_006223.3 | NP_006214.2 | 3 | 325 | |||
NR_033187.1 | 2 | 251 | ||||
GenBank mRNA | AB009690.1 | - | 3 | 246 | ||
AF143096.1 | - | 3 | 239 | |||
AK127605.1 | - | 3 | 236 | |||
AV709926.2 | - | 3 | 274 | |||
BC005234.1 | - | 3 | 244 | |||
BC070288.1 | - | 3 | 244 | |||
BC093700.1 | - | 3 | 322 | 7939545 | ||
BC104653.1 | - | 3 | 245 | |||
BC111394.1 | - | 3 | 248 | |||
BC112281.1 | - | 3 | 322 | 8327749 |