Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006223.3 | NP_006214.2 | 4 | 388 | ||
NR_033187.1 | 3 | 314 | ||||
GenBank mRNA | AB009690.1 | - | 4 | 309 | ||
AF143096.1 | - | 4 | 302 | |||
BC005234.1 | - | 4 | 307 | |||
BC070288.1 | - | 4 | 307 | |||
BC093700.1 | - | 4 | 385 | 7939545 | ||
BC104653.1 | - | 4 | 308 | |||
BC111394.1 | - | 4 | 311 | |||
BC112281.1 | - | 4 | 385 | 8327749 |
Gene Symbol: PIN4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003571.1 | 1 | 2205 | |||
GenBank mRNA | AK055299.1 | - | 1 | 2205 |