Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170747.1 | NP_001164218.1 | 3 | 300 | ||
NM_006223.3 | NP_006214.2 | 3 | 300 | |||
NR_033187.1 | 2 | 226 | ||||
GenBank mRNA | AB009690.1 | - | 3 | 221 | ||
AF143096.1 | - | 3 | 214 | |||
AK127605.1 | - | 3 | 211 | |||
AV709926.2 | - | 3 | 249 | |||
BC005234.1 | - | 3 | 219 | |||
BC070288.1 | - | 3 | 219 | |||
BC093700.1 | - | 3 | 297 | 7939545 | ||
BC104653.1 | - | 3 | 220 | |||
BC111394.1 | - | 3 | 223 | |||
BC112281.1 | - | 3 | 297 | 8327749 |
Gene Symbol: PIN4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003571.1 | 1 | 2117 | |||
GenBank mRNA | AK055299.1 | - | 1 | 2117 |