Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NUSAP1 SIRNA Assays ›
Gene Symbol:
NUSAP1
Gene Name:
nucleolar and spindle associated protein 1
Gene Aliases:
ANKT, BM037, LNP, NUSAP, PRO0310p1, Q0310, SAPL
Chromosome Location:
Chr.15: 41332694 - 41381050 on Build GRCh38
UniGene ID:
Hs.615092
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s230213
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NUSAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243142.1
NP_001230071.1
5, 6
808
NM_001243143.1
NP_001230072.1
5, 6
766
NM_001243144.1
NP_001230073.1
4, 5
742
NM_001301136.1
NP_001288065.1
5, 6
808
NM_016359.4
NP_057443.2
5, 6
811
NM_018454.7
NP_060924.4
5, 6
811
XM_005254428.2
XP_005254485.1
5, 6
683
XM_005254430.4
XP_005254487.1
5, 6
605
XM_005254431.2
XP_005254488.1
5, 6
683
XM_006720559.2
XP_006720622.1
5, 6
683
XM_006720560.3
XP_006720623.1
5, 6
779
XM_006720561.2
XP_006720624.1
5, 6
597
XM_006720562.2
XP_006720625.1
5, 6
683
XM_006720563.2
XP_006720626.1
5, 6
665
XM_017022294.1
XP_016877783.1
5, 6
605
XM_017022295.1
XP_016877784.1
5, 6
1475
GenBank mRNA
AF090915.1
-
5, 6
615
AF217513.1
-
5, 6
626
AF290612.1
-
5, 6
615
AF305711.1
-
5, 6
547
AK023483.1
-
5, 6
621
AK222819.1
-
5, 6
635
AK225703.1
-
5, 6
622
AK290879.1
-
5, 6
635
AK293168.1
-
4, 5
566
BC001308.1
-
5, 6
614
3453354
BC010838.1
-
3, 4
355
BC012887.1
-
5, 6
589
3882061
BC024772.1
-
5, 6
616
4342499
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mitotic sister chromatid segregation
mitotic cytokinesis
mitotic chromosome condensation
establishment of mitotic spindle localization
positive regulation of mitotic nuclear division
DNA binding
protein binding
microtubule binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-rhcqn:80/100.66.77.167:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline