Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128432.2 | NP_001121904.1 | 3 | 412 | ||
NM_001277225.1 | NP_001264154.1 | 5 | 1235 | |||
NM_020973.4 | NP_066024.1 | 5 | 1333 | |||
NR_102355.1 | 5 | 1333 | ||||
NR_102356.1 | 3 | 412 | ||||
NR_102357.1 | 5 | 1235 | ||||
GenBank mRNA | AB017913.1 | - | 5 | 1282 | ||
AF317840.1 | - | 5 | 1231 | |||
AF323990.1 | - | 5 | 1280 | |||
AJ278964.1 | - | 5 | 1231 | |||
AK222963.1 | - | 5 | 1280 | |||
AK292746.1 | - | 5 | 1280 | |||
AK298377.1 | - | 5 | 1235 | |||
AK313189.1 | - | 5 | 1285 | |||
BC029362.1 | - | 3 | 401 | |||
BC070188.1 | - | 3 | 392 | |||
BC101829.1 | - | 5 | 1268 | 8069335 | ||
BC109377.1 | - | 3 | 392 | |||
BC136264.1 | - | 5 | 1268 | 9020251 | ||
CB163648.1 | - | 3 | 307 |