Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOS1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126060.1 | NP_001119532.2 | 2 | 3658 | ||
NM_001164757.1 | NP_001158229.1 | 10 | 2548 | |||
NM_014697.2 | NP_055512.1 | 10 | 2563 | |||
GenBank mRNA | AB007933.2 | - | 10 | 2563 | ||
AF037070.1 | - | 2 | 1117 | |||
AY841899.1 | - | 2 | 3658 | |||
BC041382.2 | - | 10 | 2565 |