Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015077.3 | NP_055892.2 | 4, 5 | 1763 | ||
GenBank mRNA | AB011096.1 | - | 4, 5 | 1015 | ||
AJ290445.1 | - | 4, 5 | 1318 | |||
AK093615.1 | - | 1, 2 | 259 | |||
AK302038.1 | - | 5, 6 | 1092 | |||
AK314609.1 | - | 4, 5 | 1763 | |||
AY444166.1 | - | 4, 5 | 1392 | |||
BC040429.1 | - | 4, 5 | 1351 | |||
BC122860.1 | - | 5, 6 | 1071 |