Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135752.1 | NP_001129224.1 | 2 | 328 | ||
NM_001135753.1 | NP_001129225.1 | 2 | 328 | |||
NM_007265.2 | NP_009196.1 | 2 | 328 | |||
GenBank mRNA | AK022616.1 | - | 2 | 276 | ||
AK315711.1 | - | 2 | 301 | |||
BC000721.2 | - | 2 | 292 | 3504956 | ||
CR457407.1 | - | 1 | 85 | |||
D88208.1 | - | 2 | 292 | |||
DA162516.1 | - | 2 | 304 |