Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000990.4 | NP_000981.1 | 4 | 558 | ||
GenBank mRNA | AK311767.1 | - | 4 | 214 | ||
BC005326.1 | - | 1 | 214 | 4052417 | ||
BC020169.1 | - | 3 | 190 | |||
BG687024.1 | - | 248 | ||||
BQ058236.1 | - | 4 | 236 | |||
U14968.1 | - | 4 | 208 |