Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM110-MUSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198974.2 | NP_001185903.2 | 9 | 1089 | ||
GenBank mRNA | AL833659.1 | - | 3 | 4898 |
Gene Symbol: MUSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205853.3 | NP_995325.3 | 2 | 359 | ||
GenBank mRNA | AJ276556.1 | - | 2 | 204 | ||
BC130550.1 | - | 2 | 130 | 40146581 | ||
BC130552.1 | - | 2 | 128 | 40146583 | ||
BX089772.1 | - | 2 | 344 | |||
CB043913.1 | - | 2 | 369 | |||
CR990075.1 | - | 2 | 250 |