Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006945.4 | NP_008876.3 | 2 | 355 | ||
XM_017002175.1 | XP_016857664.1 | 2 | 387 | |||
GenBank mRNA | AK291317.1 | - | 2 | 357 | ||
BG183546.1 | - | 3 | 584 | |||
M21302.1 | - | 2 | 355 |