Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134432.1 | NP_001127904.1 | 5 | 1088 | ||
NM_001134433.1 | NP_001127905.1 | 5 | 1088 | |||
NM_001271650.1 | NP_001258579.1 | 6 | 1152 | |||
NM_022461.4 | NP_071906.1 | 5 | 1088 | |||
XM_005265392.2 | XP_005265449.1 | 6 | 1175 | |||
XM_011534009.2 | XP_011532311.1 | 5 | 628 | |||
XM_011534010.2 | XP_011532312.1 | 5 | 586 | |||
XM_011534011.2 | XP_011532313.1 | 5 | 585 | |||
GenBank mRNA | AK025592.1 | - | 5 | 935 | ||
AK290637.1 | - | 5 | 807 | |||
AY151386.1 | - | 4 | 556 | |||
BM546093.1 | - | 4 | 424 | |||
BX327318.2 | - | 2 | 159 | |||
BX648471.1 | - | 6 | 816 | |||
CB995808.1 | - | 5 | 616 |