Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001248.3 | NP_001239.2 | 7 | 820 | ||
NM_001291960.1 | NP_001278889.1 | 7 | 861 | |||
NM_001291961.1 | NP_001278890.1 | 7 | 820 | |||
XM_011534266.2 | XP_011532568.1 | 7 | 820 | |||
GenBank mRNA | AF034840.2 | - | 7 | 781 | ||
AF039917.1 | - | 7 | 784 | |||
AK313322.1 | - | 7 | 846 | |||
BC029869.1 | - | 6 | 724 | 5174918 |