Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDXACB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138378.2 | NP_612387.1 | 3, 4 | 859 | ||
NR_038364.1 | 2, 3 | 702 | ||||
GenBank mRNA | AK097483.1 | - | 2, 3 | 690 | ||
AK300795.1 | - | 2, 3 | 333 | |||
AK311254.1 | - | 2, 3 | 702 | |||
BC006136.1 | - | 3, 4 | 538 | |||
BC127680.1 | - | 3, 4 | 582 | 40127680 |