Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGED4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177537.2 | NP_803881.1 | 12, 13 | 2372 | ||
XM_011530822.1 | XP_011529124.1 | 12, 13 | 2329 | |||
XM_017029876.1 | XP_016885365.1 | 12, 13 | 2388 | |||
GenBank mRNA | AB040528.1 | - | 12, 13 | 2283 | ||
AB040529.1 | - | 11, 12 | 2784 | |||
AK094541.1 | - | 7, 8 | 2803 |
Gene Symbol: MAGED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098800.2 | NP_001092270.1 | 12, 13 | 2372 |