Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGED4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177537.2 | NP_803881.1 | 12 | 2359 | ||
XM_011530822.1 | XP_011529124.1 | 12 | 2316 | |||
XM_017029876.1 | XP_016885365.1 | 12 | 2375 | |||
GenBank mRNA | AB040528.1 | - | 12 | 2270 | ||
AB040529.1 | - | 11 | 2771 | |||
AK094541.1 | - | 7 | 2790 |
Gene Symbol: MAGED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098800.2 | NP_001092270.1 | 12 | 2359 |