Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PINX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284356.1 | NP_001271285.1 | 6 | 685 | ||
NM_017884.5 | NP_060354.4 | 7 | 762 | |||
GenBank mRNA | AF205718.1 | - | 6 | 674 | ||
AF418553.1 | - | 7 | 751 | |||
AK313715.1 | - | 7 | 700 | |||
AY238941.1 | - | 6 | 567 | |||
BC015479.1 | - | 7 | 708 | 3914396 | ||
BC093762.1 | - | 7 | 652 | 7939607 |