Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGACT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195087.1 | NP_001182016.1 | 3 | 1176 | ||
NM_033110.2 | NP_149101.1 | 2 | 1191 | |||
XM_005254083.2 | XP_005254140.1 | 2 | 1318 | |||
XM_011521129.2 | XP_011519431.1 | 3 | 4154 | |||
XM_017020804.1 | XP_016876293.1 | 3 | 1292 | |||
GenBank mRNA | AK091974.1 | - | 3 | 864 | ||
AK093228.1 | - | 1 | 400 | |||
AK095850.1 | - | 3 | 1169 | |||
BC001077.2 | - | 2 | 1132 | |||
BC004360.1 | - | 2 | 1132 |