Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286124.1 | NP_001273053.1 | 2 | 821 | ||
NM_001286126.1 | NP_001273055.1 | 2 | 821 | |||
NM_001286128.1 | NP_001273057.1 | 2 | 821 | |||
NM_001286129.1 | NP_001273058.1 | 2 | 821 | |||
NM_018650.4 | NP_061120.3 | 2 | 821 | |||
XM_005273134.4 | XP_005273191.1 | 2 | 688 | |||
XM_011509561.2 | XP_011507863.1 | 2 | 817 | |||
XM_011509562.2 | XP_011507864.1 | 2 | 406 | |||
XM_017001305.1 | XP_016856794.1 | 3 | 278 | |||
XM_017001306.1 | XP_016856795.1 | 2 | 290 | |||
XM_017001307.1 | XP_016856796.1 | 3 | 718 | |||
GenBank mRNA | AB040910.1 | - | 2 | 778 | ||
AF154845.1 | - | 2 | 447 | |||
AK098103.1 | - | 3 | 411 | |||
AK295506.1 | - | 2 | 319 | |||
AK314614.1 | - | 2 | 481 | |||
BC113869.1 | - | 2 | 357 | 40036731 | ||
BC114478.1 | - | 2 | 357 | 40036730 | ||
KJ534879.1 | - | 2 | 181 | |||
KJ534880.1 | - | 2 | 181 | |||
KJ534881.1 | - | 2 | 181 | |||
KJ535089.1 | - | 2 | 180 |