Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRG1 SIRNA Assays ›
Gene Symbol:
NRG1
Gene Name:
neuregulin 1
Gene Aliases:
ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, NRG1-IT2, SMDF
Chromosome Location:
Chr.8: 31639222 - 32771716 on Build GRCh38
UniGene ID:
Hs.453951
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s230507
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159995.2
NP_001153467.1
5
590
NM_001159996.2
NP_001153468.1
2
319
NM_001159999.2
NP_001153471.1
6
641
NM_001160001.2
NP_001153473.1
4
539
NM_001160002.1
NP_001153474.1
5
997
NM_001160004.2
NP_001153476.1
6
1048
NM_001160005.1
NP_001153477.1
4
946
NM_001160007.1
NP_001153479.1
4
946
NM_001160008.1
NP_001153480.1
6
1048
NM_001322197.1
NP_001309126.1
2
319
NM_001322201.1
NP_001309130.1
5
590
NM_001322202.1
NP_001309131.1
4
539
NM_001322205.1
NP_001309134.1
2
1682
NM_001322206.1
NP_001309135.1
2
1682
NM_001322207.1
NP_001309136.1
2
1682
NM_004495.3
NP_004486.2
6
1048
NM_013956.4
NP_039250.2
6
1048
NM_013957.4
NP_039251.2
6
1048
NM_013958.3
NP_039252.2
6
1048
NM_013959.3
NP_039253.1
2
1682
NM_013960.4
NP_039254.1
6
1048
NM_013962.2
NP_039256.2
4
1307
NM_013964.4
NP_039258.1
6
1048
XM_005273486.3
XP_005273543.1
2
1682
XM_005273487.3
XP_005273544.1
2
1682
XM_011544512.2
XP_011542814.1
6
831
XM_017013365.1
XP_016868854.1
6
760
XM_017013366.1
XP_016868855.1
6
760
XM_017013367.1
XP_016868856.1
4
541
XM_017013368.1
XP_016868857.1
4
878
XM_017013369.1
XP_016868858.1
2
1650
XM_017013370.1
XP_016868859.1
2
1645
XM_017013371.1
XP_016868860.1
6
760
XM_017013372.1
XP_016868861.1
6
760
GenBank mRNA
AB451331.1
-
6
531
AB451475.1
-
6
531
AF026146.1
-
6
978
AK097005.1
-
7
895
AK223435.1
-
2
1189
AK289927.1
-
2
1682
AK290730.1
-
6
947
AK293240.1
-
2
154
AK293270.1
-
2
227
AK298132.1
-
6
1048
BC006492.2
-
6
1020
BC007675.2
-
2
1339
BC064587.1
-
2
1135
BC073871.1
-
2
1303
BC144446.1
-
6
596
BC150609.1
-
6
596
9056979
BT007389.1
-
2
696
CN603652.1
-
6
559
CN603654.1
-
4
457
CN603661.1
-
2
319
CR450288.1
-
2
696
DC425692.1
-
2
239
EF372273.1
-
4
375
EF372274.1
-
4
375
EF372276.1
-
5
426
EF372277.1
-
4
375
EF517295.1
-
4
375
EF517296.1
-
6
477
EF517297.1
-
5
426
EU359055.1
-
4
426
EU363508.1
-
5
477
EU363509.1
-
4
426
EU363510.1
-
6
528
GQ983557.1
-
4
539
GQ983558.1
-
4
366
L12260.1
-
4
1335
L12261.1
-
6
924
L41827.1
-
2
1196
M94165.1
-
6
663
M94166.1
-
6
623
M94167.1
-
6
991
M94168.1
-
6
1023
S68256.1
-
6
603
U02326.1
-
6
639
U02327.1
-
4
167
U02328.1
-
4
254
U02329.1
-
4
178
U02330.1
-
5
333
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
cell morphogenesis
startle response
cardiac conduction system development
ventricular trabecula myocardium morphogenesis
cell communication
activation of transmembrane receptor protein tyrosine kinase activity
nervous system development
synapse assembly
peripheral nervous system development
locomotory behavior
regulation of blood pressure
positive regulation of cell proliferation
embryo development
neural crest cell development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
glucose transport
peptidyl-tyrosine phosphorylation
glial cell fate commitment
chemorepulsion involved in interneuron migration from the subpallium to the cortex
mammary gland development
positive regulation of myelination
activation of protein kinase B activity
intracellular signal transduction
ERBB signaling pathway
ERBB2 signaling pathway
wound healing
negative regulation of protein catabolic process
myelination
regulation of protein homodimerization activity
regulation of protein heterodimerization activity
positive regulation of GTPase activity
neurotransmitter receptor metabolic process
positive regulation of axon extension
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of Ras protein signal transduction
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
neuron fate commitment
oligodendrocyte differentiation
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of secretion
positive regulation of striated muscle cell differentiation
positive regulation of protein kinase B signaling
cardiac muscle cell differentiation
cardiac muscle cell myoblast differentiation
endocardial cell differentiation
positive regulation of dendritic spine development
positive regulation of calcineurin-NFAT signaling cascade
positive regulation of protein targeting to mitochondrion
positive regulation of protein localization to cell surface
regulation of cell motility
positive regulation of cardiac muscle cell differentiation
negative regulation of neuron migration
transcription cofactor activity
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
cytokine activity
ErbB-2 class receptor binding
integrin binding
growth factor activity
transmembrane receptor protein tyrosine kinase activator activity
receptor tyrosine kinase binding
ErbB-3 class receptor binding
chemorepellent activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0