Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290263.1 | NP_001277192.1 | 52 | 5995 | ||
NM_014689.2 | NP_055504.2 | 52 | 6138 | |||
XM_005246673.4 | XP_005246730.1 | 52 | 5997 | |||
XM_006712617.3 | XP_006712680.1 | 52 | 6162 | |||
XM_017004441.1 | XP_016859930.1 | 52 | 6165 | |||
XM_017004442.1 | XP_016859931.1 | 52 | 6141 | |||
XM_017004443.1 | XP_016859932.1 | 52 | 6214 | |||
XM_017004444.1 | XP_016859933.1 | 51 | 6121 | |||
XM_017004445.1 | XP_016859934.1 | 52 | 5995 | |||
XM_017004446.1 | XP_016859935.1 | 51 | 6019 | |||
XM_017004447.1 | XP_016859936.1 | 52 | 6137 | |||
GenBank mRNA | AB074179.1 | - | 7 | 728 | ||
AF250396.1 | - | 12 | 1414 | |||
AK001253.1 | - | 7 | 738 | |||
AK293847.1 | - | 18 | 2064 | |||
AK293879.1 | - | 23 | 2451 | |||
AK307914.1 | - | 12 | 1499 | |||
BC015018.1 | - | 10 | 1105 | |||
CR749492.1 | - | 27 | 2983 | |||
EU236710.2 | - | 52 | 5992 |