Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290263.1 | NP_001277192.1 | 26 | 3049 | ||
NM_014689.2 | NP_055504.2 | 26 | 3192 | |||
XM_005246673.4 | XP_005246730.1 | 26 | 3051 | |||
XM_006712617.3 | XP_006712680.1 | 26 | 3216 | |||
XM_017004441.1 | XP_016859930.1 | 26 | 3219 | |||
XM_017004442.1 | XP_016859931.1 | 26 | 3222 | |||
XM_017004443.1 | XP_016859932.1 | 26 | 3268 | |||
XM_017004444.1 | XP_016859933.1 | 26 | 3220 | |||
XM_017004445.1 | XP_016859934.1 | 26 | 3049 | |||
XM_017004446.1 | XP_016859935.1 | 26 | 3220 | |||
XM_017004447.1 | XP_016859936.1 | 26 | 3218 | |||
GenBank mRNA | CR749492.1 | - | 1 | 37 | ||
EU236710.2 | - | 26 | 3046 |