Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFB1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016020.3 | NP_057104.2 | 7 | 946 | ||
XM_005267006.4 | XP_005267063.1 | 7 | 1124 | |||
XM_011535873.2 | XP_011534175.1 | 6 | 1666 | |||
XM_017010917.1 | XP_016866406.1 | 7 | 860 | |||
GenBank mRNA | AF151833.1 | - | 7 | 963 | ||
AK289484.1 | - | 7 | 960 | |||
BC005183.1 | - | 7 | 944 | |||
BC017788.1 | - | 7 | 946 | 4672440 |