Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFB1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016020.3 | NP_057104.2 | 3 | 391 | ||
XM_005267006.4 | XP_005267063.1 | 3 | 569 | |||
XM_011535870.1 | XP_011534172.1 | 3 | 405 | |||
XM_011535871.1 | XP_011534173.1 | 3 | 405 | |||
XM_011535872.2 | XP_011534174.1 | 3 | 405 | |||
XM_011535873.2 | XP_011534175.1 | 2 | 1111 | |||
XM_017010917.1 | XP_016866406.1 | 3 | 305 | |||
GenBank mRNA | AF151833.1 | - | 3 | 408 | ||
AK289484.1 | - | 3 | 405 | |||
BC005183.1 | - | 3 | 389 | |||
BC017788.1 | - | 3 | 391 | 4672440 | ||
BU849912.1 | - | 3 | 400 |