Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001560.2 | NP_001001560.1 | 1, 2 | 434 | ||
NM_001172687.1 | NP_001166158.1 | 1, 2 | 434 | |||
NM_013365.4 | NP_037497.1 | 1, 2 | 434 | |||
XM_005261517.3 | XP_005261574.1 | 2, 3 | 608 | |||
XM_011530122.1 | XP_011528424.1 | 2, 3 | 541 | |||
XM_011530124.1 | XP_011528426.1 | 2, 3 | 608 | |||
XM_017028760.1 | XP_016884249.1 | 2, 3 | 608 | |||
XM_017028762.1 | XP_016884251.1 | 2, 3 | 608 | |||
XM_017028763.1 | XP_016884252.1 | 1, 2 | 74 | |||
GenBank mRNA | AF190862.1 | - | 1, 2 | 62 | ||
AF218584.1 | - | 1, 2 | 51 | |||
AF233521.1 | - | 1, 2 | 69 | |||
AK075256.1 | - | 1, 2 | 412 | |||
AK225157.1 | - | 1, 2 | 68 | |||
AK290548.1 | - | 1, 2 | 73 | |||
BC010917.1 | - | 1, 2 | 78 | |||
BC029388.1 | - | 1, 2 | 73 | |||
BC044629.1 | - | 1, 2 | 63 | 5796877 | ||
CR456493.1 | - | 1, 2 | 183 |