Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017875.2 | NP_060345.2 | 6 | 1181 | ||
XM_006713214.1 | XP_006713277.1 | 7 | 1144 | |||
XM_011533869.2 | XP_011532171.1 | 7 | 2044 | |||
GenBank mRNA | AK000558.1 | - | 6 | 1157 | ||
AK026356.1 | - | 2 | 174 | |||
AK225388.1 | - | 6 | 1133 | |||
BC013194.1 | - | 6 | 842 | 3918744 | ||
CR457242.1 | - | 6 | 780 |