Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297752.1 | NP_001284681.1 | 6 | 1465 | ||
NM_181845.1 | NP_862828.1 | 7 | 1520 | |||
XM_005258784.3 | XP_005258841.1 | 7 | 1633 | |||
XM_005258785.3 | XP_005258842.1 | 6 | 1427 | |||
XM_005258786.3 | XP_005258843.1 | 7 | 1592 | |||
XM_017026628.1 | XP_016882117.1 | 8 | 1714 | |||
XM_017026629.1 | XP_016882118.1 | 8 | 1755 | |||
XM_017026630.1 | XP_016882119.1 | 8 | 1718 | |||
XM_017026631.1 | XP_016882120.1 | 8 | 1585 | |||
XM_017026632.1 | XP_016882121.1 | 8 | 1761 | |||
XM_017026633.1 | XP_016882122.1 | 8 | 1710 | |||
XM_017026634.1 | XP_016882123.1 | 7 | 1463 | |||
XM_017026635.1 | XP_016882124.1 | 6 | 1528 | |||
XM_017026636.1 | XP_016882125.1 | 2 | 7233 | |||
XM_017026637.1 | XP_016882126.1 | 6 | 1506 | |||
XM_017026638.1 | XP_016882127.1 | 5 | 1300 | |||
XM_017026639.1 | XP_016882128.1 | 7 | 1628 | |||
GenBank mRNA | AK098175.1 | - | 6 | 1377 | ||
AK294852.1 | - | 7 | 1543 | |||
AY500359.1 | - | 1 | 739 | |||
AY500364.1 | - | 1 | 611 |