Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100915.1 | NP_001094385.1 | 13 | 2401 | ||
GenBank mRNA | AK302567.1 | - | 12 | 3619 | ||
AK302748.1 | - | 13 | 2385 | |||
BC070103.1 | - | 13 | 4182 | |||
BC094790.1 | - | 11 | 4348 | |||
BC117335.1 | - | 13 | 2401 | 40125886 |