Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100595.1 | NP_001094065.1 | 3 | 665 | ||
NM_182620.3 | NP_872426.1 | 4 | 667 | |||
XM_011524745.2 | XP_011523047.1 | 4 | 501 | |||
GenBank mRNA | AK022820.1 | - | 3 | 222 | ||
AK056473.1 | - | 4 | 419 | |||
BC017873.1 | - | 4 | 428 | 4685972 | ||
BC106003.1 | - | 4 | 392 | 6143473 | ||
DA133216.1 | - | 3 | 403 | |||
DB158057.1 | - | 2 | 403 |