Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142272.1 | NP_001135744.1 | 9 | 1120 | ||
NM_014700.3 | NP_055515.1 | 11 | 2166 | |||
XM_005255713.3 | XP_005255770.1 | 12 | 2336 | |||
XM_005255714.1 | XP_005255771.1 | 10 | 1181 | |||
XM_005255715.4 | XP_005255772.1 | 10 | 1987 | |||
XM_011522764.2 | XP_011521066.1 | 10 | 1213 | |||
XM_011522765.2 | XP_011521067.1 | 10 | 1039 | |||
GenBank mRNA | AB014565.1 | - | 11 | 2214 | ||
AF395731.1 | - | 11 | 2214 | |||
AK293644.1 | - | 9 | 1035 | |||
AK303061.1 | - | 9 | 1124 | |||
AY130007.1 | - | 3 | 235 | |||
BC051360.1 | - | 11 | 2165 | 6646253 |