Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142294.1 | NP_001135766.1 | 2 | 269 | ||
NM_001142295.1 | NP_001135767.1 | 2 | 393 | |||
NM_001142296.1 | NP_001135768.1 | 3 | 445 | |||
NM_015087.4 | NP_055902.1 | 2 | 462 | |||
XM_005266313.4 | XP_005266370.1 | 2 | 387 | |||
XM_005266314.3 | XP_005266371.1 | 3 | 436 | |||
XM_005266315.3 | XP_005266372.1 | 3 | 290 | |||
XM_005266317.3 | XP_005266374.1 | 2 | 220 | |||
XM_011535012.2 | XP_011533314.1 | 2 | 373 | |||
GenBank mRNA | AB011182.1 | - | 2 | 236 | ||
AK001949.1 | - | 2 | 226 | |||
AK057764.1 | - | 2 | 242 | |||
AK291724.1 | - | 2 | 313 | |||
AY038359.1 | - | 1 | 175 | |||
AY123329.1 | - | 2 | 236 | |||
BC047083.1 | - | 2 | 269 | 5313378 | ||
BX648004.1 | - | 2 | 358 | |||
DA347686.1 | - | 2 | 350 | |||
DA507821.1 | - | 2 | 373 | |||
DA710623.1 | - | 2 | 437 | |||
KU178563.1 | - | 1 | 175 | |||
KU178564.1 | - | 1 | 175 |