Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024843.1 | NP_001020014.1 | 12 | 1423 | ||
NM_001162501.1 | NP_001155973.1 | 10 | 3573 | |||
NM_015088.2 | NP_055903.2 | 9 | 3414 | |||
GenBank mRNA | AB029016.2 | - | 9 | 3397 | ||
AK294519.1 | - | 10 | 3564 | |||
AK304087.1 | - | 6 | 899 | |||
BC028626.2 | - | 12 | 1356 | 4826787 | ||
BX647630.1 | - | 6 | 2616 |